Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-TSC2 [p Thr1462], Polyclonal, Novus Biologicals™

Manufacturer:  Novus Biologicals NBP141926100UG

Catalog No. NBP14192610

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

TSC2 (p Thr1462)
1 mg/mL
Western Blot 1:500-1:1000, ELISA 1:4000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:50-1:100
P49815
LAMFLJ43106, TSC4, tuberin, tuberous sclerosis 2, Tuberous sclerosis 2 protein
Rabbit
IgG
Immunogen affinity purified
RUO
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Polyclonal
Human, Mouse, Rat
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Unconjugated
PBS (without Mg2+ and Ca2+), pH 7.4, 150mM NaCl, and 50% glycerol. with 0.02% Sodium Azide
Affinity Purified
TSC2
synthesized phosphopeptide derived from human Tsc2 around the phosphorylation site of threonine 1462(G-Y-TP-I-S).
200 kDa
100 ug
Apoptosis, Autophagy, Cancer, Cell Cycle and Replication, Cytoskeleton Markers, Hypoxia, mTOR Pathway, Phospho Specific, Signal Transduction, Tumor Suppressors
Primary
7249
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit