Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

TSC2 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic peptide of TSC2.

Manufacturer:  Abnova Corporation PAB13146

Catalog No. 89-116-393


Add to cart

Description

Description

Mutations in this gene lead to tuberous sclerosis complex. Its gene product is believed to be a tumor suppressor and is able to stimulate specific GTPases. The protein associates with hamartin in a cytosolic complex, possibly acting as a chaperone for hamartin. Alternative splicing results in multiple transcript variants encoding different isoforms. [provided by RefSeq

Specifications

Specifications

TSC2
Unconjugated
Western Blot (1 ug/mL) The optimal working dilution should be determined by the end user.
TSC2
FLJ43106|LAM|TSC4
Rabbit
100 ug
Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human, Murine
Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of TSC2.
In PBS (0.02% sodium azide)
Liquid
TSC2
A synthetic peptide corresponding to C-terminus of human TSC2.
RUO
Primary
7249
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only