Learn More
TTBK2, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of TTBK2.
Supplier:  Abnova Corporation PAB27186
Catalog No. 89-128-838
Add to Cart
Description
Description
This gene encodes a serine-threonine kinase that putatively phosphorylates tau and tubulin proteins. Mutations in this gene cause spinocerebellar ataxia type 11 (SCA11); a neurodegenerative disease characterized by progressive ataxia and atrophy of the cerebellum and brainstem. [provided by RefSeq
Specifications
Specifications
TTBK2
Polyclonal
Unconjugated
Immunohistochemistry (5 ug/mL) The optimal working dilution should be determined by the end user.
TTBK2
TTBK2
A synthetic peptide corresponding to 18 amino acids at C-terminus of human TTBK2.
100 μg
Primary
TTBK2 antibody is human specific. At least four isoforms of TTBK2 are known to exist; this antibody will detect only the two largest isoforms.
Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
IgG
ELISA, Immunohistochemistry
1 mg/mL
Rabbit polyclonal antibody raised against synthetic peptide of TTBK2.
In PBS (0.02% sodium azide)
KIAA0847/SCA11/TTBK
Rabbit
Peptide affinity purification
RUO
146057
Human
Liquid
Product Suggestions
Product Suggestions