Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
TUBB3 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of TUBB3.
Supplier: Abnova Corporation PAB24742
Specifications
TUBB3 | |
Polyclonal | |
Unconjugated | |
Western Blot (1:5000-1:10000) The optimal working dilution should be determined by the end user. | |
TUBB3 | |
TUBB3 | |
A synthetic peptide corresponding to TUBB3. | |
100 μL | |
Primary | |
TUBB3 polyclonal antibody detects endogenous levels of TUBB3 protein. | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Western Blot | |
1 mg/mL | |
Rabbit polyclonal antibody raised against synthetic peptide of TUBB3. | |
In PBS, pH 7.2 (0.05% sodium azide) | |
MC1R/TUBB4/beta-4 | |
Rabbit | |
Antigen affinity purification | |
RUO | |
10381 | |
Human, Mouse, Rat | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction