Learn More
UNC119, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of UNC119.
Supplier:  Abnova Corporation PAB5396
Catalog No. 89-131-843
Add to Cart
Description
Description
This gene is specifically expressed in the photoreceptors in the retina. The encoded product shares strong homology with the C. elegans unc119 protein and it can functionally complement the C. elegans unc119 mutation. It has been localized to the photoreceptor synapses in the outer plexiform layer of the retina, and suggested to play a role in the mechanism of photoreceptor neurotransmitter release through the synaptic vesicle cycle. Two transcript variants encoding different isoforms have been described for this gene. [provided by RefSeq
Specifications
Specifications
UNC119
Polyclonal
Rabbit polyclonal antibody raised against synthetic peptide of UNC119.
In PBS, pH 7.2 (50% glycerol, 0.01% sodium azide)
HRG4
Rabbit
100 μg
Primary
Human, Mouse, Rat
Liquid
ELISA, Western Blot
Unconjugated
The optimal working dilution should be determined by the end user.
UNC119
UNC119
A synthetic peptide corresponding to N-terminus of human UNC119.
RUO
9094
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Recommendations
Recommendations