Learn More
UPK3A, Rabbit, Clone: SP73, Abnova™
Rabbit monoclonal antibody raised against synthetic peptide of UPK3A.
Manufacturer:  Abnova Corporation MAB10029
Catalog No. 89-108-343
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Uroplakins (UPK) are integral membrane proteins that form 2-dimensional crystalline arrays termed urothelial plaques that cover more than 90% of the apical urothelial surface. The asymmetric unit membrane (AUM) forms the apical plaques of urothelium and is believed to strengthen the urothelial apical surface and prevent the cells from rupturing during bladder distention. The 4 major conserved integral membrane proteins of the AUM are UPK1A (MIM 611557), UPK1B (MIM 602380), UPK2 (MIM 611558), and UPK3A.[supplied by OMIM
Specifications
Specifications
UPK3A
SP73
Rabbit monoclonal antibody raised against synthetic peptide of UPK3A.
In 10mM PBS, pH 7.4 (BSA, 0.09% sodium azide)
Liquid
UPK3A
A synthetic peptide corresponding to C-terminus of human UPK3A.
100 uL
Store at 4°C. Do not freeze.
Monoclonal
Human
Immunohistochemistry (PFA fixed)
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100) The optimal working dilution should be determined by the end user.
UPK3A
MGC119178/UPIII/UPIIIA/UPK3
Rabbit
Affinity purification
RUO
Primary
7380