Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
USP20 Rabbit anti-Human, Monkey, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of USP20.
Manufacturer: Abnova Corporation PAB16496
Specifications
USP20 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of USP20. | |
In PBS (0.09% sodium azide) | |
KIAA1003/LSFR3A/VDU2 | |
Rabbit | |
Immunoaffinity purification | |
RUO | |
10868 | |
Human, Monkey |
Immunohistochemistry (PFA fixed) | |
Unconjugated | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (10 ug/mL) The optimal working dilution should be determined by the end user. | |
USP20 | |
USP20 | |
A synthetic peptide (conjugated with KLH) corresponding to human USP20. | |
50 μg | |
Primary | |
Internal domain of human. | |
Liquid |
Safety and Handling
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only