Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
USP5, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of USP5.
Supplier: Abnova Corporation PAB4182
Specifications
USP5 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of USP5. | |
In PBS (0.09% sodium azide) | |
ISOT | |
Rabbit | |
Protein G purification | |
RUO | |
8078 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Western Blot | |
Unconjugated | |
ELISA (1:1000) Western Blot (1:100-500) The optimal working dilution should be determined by the end user. | |
USP5 | |
USP5 | |
A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human USP5. | |
400 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction