Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VHL, Rabbit, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic peptide of VHL.

Manufacturer:  Abnova Corporation PAB12546

Catalog No. 89-115-915


Add to cart

Description

Description

Specifications

Specifications

VHL
Unconjugated
Western Blot (1:500-1:1000) The optimal working dilution should be determined by the end user.
Liquid
VHL
A synthetic peptide corresponding to human and mouse VHL/Vhl.
RUO
Primary
7428|22346
Human, Mouse
Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of VHL.
In Tris-citrate/phosphate buffer, pH 7.0-8.0 (0.09% sodium azide)
HRCA1|RCA1|VHL1
Rabbit
100 uL
Store at 4°C. Do not freeze.
Polyclonal
This antibody is specific to VHL protein.
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only