Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

VSV-G Epitope tag Rabbit, Polyclonal Antibody, Abnova™

Catalog No. 89138283 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-138-283 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-138-283 Supplier Abnova Corporation Supplier No. PAB17072
Add to Cart
Add to Cart
Request a Quote

Rabbit polyclonal antibody raised against synthetic peptides of VSV-G Epitope tag.

Sequence: YTDIEMNRLGK

Specifications

Antigen VSV-G Epitope tag
Applications ELISA, Immunocytochemistry, Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Description Rabbit polyclonal antibody raised against synthetic peptides of VSV-G Epitope tag.
Dilution ELISA (2-10 ug/mL) Immunohistochemistry (0.5-5 ug/mL) Immunocytochemistry (0.5-5 ug/mL) The optimal working dilution should be determined by the end user.
Formulation In PBS
Host Species Rabbit
Immunogen A synthetic peptide (conjugated with KLH) corresponding to VSV-G tag.
Quantity 1 mg
Regulatory Status RUO
Primary or Secondary Primary
Content And Storage Store at 4°C.
Form Liquid
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.