Learn More
WEE1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of WEE1.
Supplier: Abnova Corporation PAB3321
Description
This gene encodes a nuclear protein, which is a tyrosine kinase belonging to the Ser/Thr family of protein kinases. This protein catalyzes the inhibitory tyrosine phosphorylation of CDC2/cyclin B kinase, and appears to coordinate the transition between DNA replication and mitosis by protecting the nucleus from cytoplasmically activated CDC2 kinase. [provided by RefSeq
Specifications
WEE1 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of WEE1. | |
In PBS (0.09% sodium azide) | |
DKFZp686I18166/FLJ16446/WEE1A/WEE1hu | |
Rabbit | |
Protein G purification | |
RUO | |
7465 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
ELISA, Immunohistochemistry, Western Blot | |
Unconjugated | |
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:50-100) Western Blot (1:1000) The optimal working dilution should be determined by the end user. | |
WEE1 | |
WEE1 | |
A synthetic peptide (conjugated with KLH) corresponding to C-terminus of human WEE1. | |
400 μL | |
Primary | |
Human | |
Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only