Learn More
West Nile Virus Envelope Rabbit anti-Viruses, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of West Nile Virus Envelope.
Manufacturer:  Abnova Corporation PAB13413
Catalog No. 89-137-595
Add to cart
Description
Description
Specifications
Specifications
West Nile Virus Envelope
Unconjugated
ELISA (1 ug/mL) The optimal working dilution should be determined by the end user.
Liquid
A synthetic peptide corresponding to C-terminus 14 amino acids of West Nile virus Envelope.
RUO
Primary
Virus
ELISA
Rabbit polyclonal antibody raised against synthetic peptide of West Nile Virus Envelope.
In PBS (0.02% sodium azide)
Rabbit
100 ug
Store at 4°C for three months. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal