Learn More
WNT1 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant WNT1.
Manufacturer: Abnova Corporation PAB18007
Description
The WNT gene family consists of structurally related genes which encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. This gene is a member of the WNT gene family. It is very conserved in evolution, and the protein encoded by this gene is known to be 98% identical to the mouse Wnt1 protein at the amino acid level. The studies in mouse indicate that the Wnt1 protein functions in the induction of the mesencephalon and cerebellum. This gene was originally considered as a candidate gene for Joubert syndrome, an autosomal recessive disorder with cerebellar hypoplasia as a leading feature. However, further studies suggested that the gene mutations might not have a significant role in Joubert syndrome. This gene is clustered with another family member, WNT10B, in the chromosome 12q13 region. [provided by RefSeq
Specifications
WNT1 | |
Polyclonal | |
Rabbit polyclonal antibody raised against recombinant WNT1. | |
In PBS pH 7.2 (30% glycerol, 0.5% BSA, 0.01% thimerosal) | |
INT1 | |
Rabbit | |
100 μg | |
Primary | |
Recombinant human Wnt-1 can be used as a positive control. | |
Liquid |
Western Blot | |
Unconjugated | |
Western Blot (0.5-4 ug/mL) The optimal working dilution should be determined by the end user. | |
WNT1 | |
WNT1 | |
Recombinant protein corresponding to human WNT1. | |
RUO | |
7471 | |
Human |
Safety and Handling
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only