Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

XRCC5 Rabbit anti-Human, Mouse, Rat, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic peptide of Xrcc5.

Manufacturer:  Abnova Corporation PAB12580

Catalog No. 89-115-927


Add to cart

Description

Description

Sequence: FSKVDEEQMKYKSEGK
Specifications

Specifications

XRCC5
Unconjugated
The optimal working dilution should be determined by the end user.
Xrcc5
AI314015|Ku80|Ku86
Rabbit
100 ug
Store at 4°C for short term. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Reacts with residues 323-338 [FSKVDEEQMKYKSEGK] of the 80 KDa Ku80 protein. Sequence is 100% conserved between human, mouse rat and hamster.
Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of Xrcc5.
In PBS (0.02% sodium azide)
Liquid
Xrcc5
A synthetic peptide corresponding to amino acids 323-338 of Xrcc5.
RUO
Primary
22596
Human, Murine, Rat
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only