Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZBED3 Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ZBED3.
Manufacturer: Abnova Corporation PAB16725
Specifications
ZBED3 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of ZBED3. | |
In PBS (0.02% sodium azide) | |
MGC15435 | |
Rabbit | |
100 μg | |
Primary | |
Human |
ELISA, Immunocytochemistry, Western Blot | |
Unconjugated | |
Western Blot (1-2 ug/mL) The optimal working dilution should be determined by the end user. | |
ZBED3 | |
ZBED3 | |
A synthetic peptide corresponding to C-terminus 15 amino acids of human ZBED3. | |
RUO | |
84327 | |
Liquid |
Safety and Handling
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only