Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZBED5, Rabbit, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against partial recombinant ZBED5.
Supplier: Abnova Corporation PAB18531
Specifications
ZBED5 | |
Polyclonal | |
Rabbit polyclonal antibody raised against partial recombinant ZBED5. | |
In buffer containing 0.02% sodium azide | |
Rabbit | |
Protein A purification | |
RUO | |
58386 | |
Human | |
Liquid |
ELISA, Immunohistochemistry (PFA fixed), Western Blot | |
Unconjugated | |
Western Blot (1:500-1:1000) Immunohistochemistry (1:100-1:500) ELISA (1:20000-1:80000) The optimal working dilution should be determined by the end user. | |
BC047754 | |
Recombinant protein corresponding to amino acids 224-474 of human ZBED5. | |
100 μg | |
Primary | |
This antibody is specific to ZBED5. | |
Store at 4°C for three months. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction