Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZC3H12D Rabbit anti-Human, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against recombinant ZC3H12D.
Supplier: Abnova Corporation PAB23080
Description
Sequence: GYDREDVLRVLGKLGEGALVNDVLQELIRTGSRPGALEHPAAPRLVPRGSCGVPDSAQRGPGTALEEDFRTLASpecifications
ZC3H12D | |
Polyclonal | |
Rabbit polyclonal antibody raised against recombinant ZC3H12D. | |
In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide) | |
C6orf95/FLJ46041/MCPIP4/dJ281H8.1/p34 | |
Rabbit | |
Antigen affinity purification | |
RUO | |
340152 | |
Store at 4°C. For long term storage store at -20°C. Aliquot to avoid repeated freezing and thawing. |
|
IgG |
Immunohistochemistry, Immunohistochemistry (Formalin/Paraffin) | |
Unconjugated | |
Immunohistochemistry (1:50-1:200) The optimal working dilution should be determined by the end user. | |
ZC3H12D | |
ZC3H12D | |
Recombinant protein corresponding to amino acids of human ZC3H12D. | |
100 μL | |
Primary | |
Human | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction