Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZDHHC6, Rabbit, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against recombinant ZDHHC6.

Manufacturer:  Abnova Corporation PAB20715

Catalog No. 89-122-215


Add to cart

Description

Description

Sequence: QLYHRLSFGWNTVKIDMSAARRDPLPIVPFGLAAF
Specifications

Specifications

ZDHHC6
Unconjugated
Immunohistochemistry (1:200-1:500) Western Blot (1:250-1:500) The optimal working dilution should be determined by the end user.
ZDHHC6
FLJ21952|ZNF376
Rabbit
IgG
100 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
Immunohistochemistry (PFA fixed), Western Blot
Rabbit polyclonal antibody raised against recombinant ZDHHC6.
In PBS, pH 7.5 (40% glycerol, 0.02% sodium azide)
Liquid
ZDHHC6
Recombinant protein corresponding to amino acids of human ZDHHC6.
Antigen affinity purification
RUO
Primary
64429
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only