Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZFP109 Rabbit anti-Human, Mouse, Rat, Porcine, Bovine, Canine, Equine, Guinea Pig, Rabbit, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236484

Catalog No. NBP236484


Add to cart

Description

Description

ZFP109 Polyclonal antibody specifically detects ZFP109 in Human, Mouse, Rat, Porcine, Bovine, Canine, Equine, Guinea Pig, Rabbit samples. It is validated for Western Blot.
Specifications

Specifications

ZFP109
Unconjugated
PBS buffer and 2% sucrose with 0.09% Sodium Azide
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
7761
Western Blot
Western Blot
Affinity Purified
ARP38428_P050
73 kDa
100 ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Bovine, Canine, Equine, Guinea Pig, Human, Mouse, Porcine, Rabbit, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit