Learn More
ZFP36L2 Rabbit anti-Human, Mouse, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ZFP36L2.
Manufacturer: Abnova Corporation PAB17366
Description
This gene is a member of the TIS11 family of early response genes. Family members are induced by various agonists such as the phorbol ester TPA and the polypeptide mitogen EGF. The encoded protein contains a distinguishing putative zinc finger domain with a repeating cys-his motif. This putative nuclear transcription factor most likely functions in regulating the response to growth factors. [provided by RefSeq
Specifications
ZFP36L2 | |
Polyclonal | |
Rabbit polyclonal antibody raised against synthetic peptide of ZFP36L2. | |
In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol) | |
BRF2/ERF-2/ERF2/RNF162C/TIS11D | |
Rabbit | |
100 μg | |
Primary | |
This antibody detects endogenous levels of total ZFP36L2 protein. | |
Liquid |
Western Blot | |
Unconjugated | |
Western Blot (1:500-1:1000) ELISA (1:20000) The optimal working dilution should be determined by the end user. | |
ZFP36L2 | |
ZFP36L2 | |
A synthetic peptide corresponding to internal of human ZFP36L2. | |
RUO | |
678 | |
Human, Mouse |
Safety and Handling
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
For Research Use Only