Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit anti-ZFYVE16, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP236538

Catalog No. NBP236538


Add to cart

Description

Description

ZFYVE16 Polyclonal antibody specifically detects ZFYVE16 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications

Specifications

ZFYVE16
1.0 mg/mL
Western Blot 1:2000 - 1:10000, Immunoprecipitation 2 - 10 ug/mg
NP_001098721.1
DKFZp686E13162, ENDOFIN, endosome-associated FYVE-domain protein, KIAA0305Endosome-associated FYVE domain protein, zinc finger FYVE domain-containing protein 16, zinc finger, FYVE domain containing 16
Rabbit
IgG
100 ul
Store at 4C. Do not freeze.
Polyclonal
Human
Western Blot, Immunoprecipitation
Unconjugated
Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Affinity Purified
ZFYVE16
The immunogen recognized by the antibody maps to a region between residue 450 to 500 of human Endosome-Associated FYVE Domain Protein using the numbering given in entry NP_001098721.1.
Immunogen affinity purified
RUO
Primary
9765
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit