Learn More
ZMPSTE24 Rabbit anti-Human, Mouse, Primates, Zebra fish, Biotin, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ZMPSTE24.
Manufacturer:  Abnova Corporation PAB15059
Catalog No. 89-117-425
Add to cart
Description
Description
This gene encodes a member of the peptidase M48A family. The encoded protein is a zinc metalloproteinase involved in the two step post-translational proteolytic cleavage of carboxy terminal residues of farnesylated prelamin A to form mature lamin A. Mutations in this gene have been associated with mandibuloacral dysplasia and restrictive dermopathy. [provided by RefSeq
Specifications
Specifications
ZMPSTE24
Biotin
Western Blot (2 ug/mL) The optimal working dilution should be determined by the end user.
ZMPSTE24
FACE-1|FACE1|FLJ14968|STE24|Ste24p
Rabbit
100 uL
Store at 4°C. Do not freeze.
Polyclonal
This is specific to ZMPSTE24. By Western Blot, a band is seen at ~54 KDa representing ZMPSTE24 protein.
Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of ZMPSTE24.
In PBS
Liquid
ZMPSTE24
A synthetic peptide corresponding to amino acids 1-100 of human ZMPSTE24.
RUO
Primary
10269
Human, Murine, Primate, Zebrafish