Learn More
ZMPSTE24 Rabbit anti-Human, DyLight 649, Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against synthetic peptide of ZMPSTE24.
Manufacturer:  Abnova Corporation PAB15054
Catalog No. 89-117-420
Add to cart
Description
Description
This gene encodes a member of the peptidase M48A family. The encoded protein is a zinc metalloproteinase involved in the two step post-translational proteolytic cleavage of carboxy terminal residues of farnesylated prelamin A to form mature lamin A. Mutations in this gene have been associated with mandibuloacral dysplasia and restrictive dermopathy. [provided by RefSeq
Specifications
Specifications
ZMPSTE24
DyLight 649
Western Blot (2 ug/ mL) The optimal working dilution should be determined by the end user.
ZMPSTE24
FACE-1|FACE1|FLJ14968|STE24|Ste24p
Rabbit
100 uL
Store at 4°C. Do not freeze.
Polyclonal
This is specific to ZMPSTE24. By Western blot a band is seen at ~54 KDa, representing ZMPSTE24 protein.
Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of ZMPSTE24.
In 50mM sodium borate
Liquid
ZMPSTE24
A synthetic peptide corresponding to amino acids 400-475 of human ZMPSTE24.
RUO
Primary
10269
Human