Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF682, Rabbit, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic peptide of ZNF682.

Manufacturer:  Abnova Corporation PAB17350

Catalog No. 89-119-006


Add to cart

Description

Description

Specifications

Specifications

ZNF682
Unconjugated
Western Blot (1:500-1:1000) ELISA (1:10000) The optimal working dilution should be determined by the end user.
ZNF682
BC39498_3|FLJ90362
Rabbit
100 ug
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
This antibody detects endogenous levels of total ZNF682 protein.
Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of ZNF682.
In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol)
Liquid
ZNF682
A synthetic peptide corresponding to internal of human ZNF682.
RUO
Primary
91120
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only