Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF76, Rabbit, Polyclonal Antibody, Abnova™

Rabbit polyclonal antibody raised against synthetic peptide of ZNF76.

Manufacturer:  Abnova Corporation PAB17363

Catalog No. 89-119-019


Add to cart

Description

Description

Specifications

Specifications

ZNF76
Unconjugated
Immunohistochemistry (1:50-1:100) ELISA (1:10000) The optimal working dilution should be determined by the end user.
ZNF76
D6S229E|ZNF523|Zfp523
Rabbit
100 ug
Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
This antibody detects endogenous levels of total ZNF76 protein.
Immunohistochemistry (PFA fixed)
Rabbit polyclonal antibody raised against synthetic peptide of ZNF76.
In PBS, pH 7.4 (150mM NaCl, 0.02% sodium azide, 50% glycerol)
Liquid
ZNF76
A synthetic peptide corresponding to internal of human ZNF76.
RUO
Primary
7629
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit

For Research Use Only