Learn More
Aviva Systems Biology Corp RABBIT ANTIBODY 50UG/50UL
Supplier:  Aviva Systems Biology Corp ARP50941_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LOC730950. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human LOC730950. Alternate names: -ApplicationWB. Species reactivity: Guinea Pig, Human, Mouse, Rabbit, Zebrafish
Catalog No. 50-014-60471
Item Discontinued This item has been discontinued and is no longer available. Please reach out to your sales representative.