Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Rabbit C-Peptide ELISA Kit (Colorimetric)

Assay has high sensitivity and specificity for detection of rabbit C-Peptide

$732.50

Specifications

Species Rabbit
Sensitivity 0.3ng/mL
Sample Type serum, plasma
Research Discipline Lipid and Metabolism
Specificity For detection of rabbit C-Peptide
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB599591KT View Documents Novus Biologicals™
NBP2599591KIT
1 Kit Each for $732.50
Add to cart
 
Description

Description

The Rabbit C-Peptide ELISA Kit (Colorimetric) measures rabbit C-Peptide in serum, plasma.
Specifications

Specifications

Rabbit
serum, plasma
For detection of rabbit C-Peptide
Store the unopened product at 2 to 8°C Protect from light. Do not use past expiration date.
ELISA
INS
0.5ng/mL to 15ng/mL
Assay plate, Adhesive Strip (For 96 wells), HRP-conjugate, Wash Buffer (20 x concentrate), Substrate B, Standard, Stop Solution, Substrate A, Instruction manual
0.3ng/mL
Lipid and Metabolism
IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin
C-Peptide
3630
ELISA Kit (Colorimetric)
50 - 100μL

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit