Learn More
Kingfisher Biotech Inc Rabbit CXCL10 (IP-10) ELISA
SDP
Supplier:  Kingfisher Biotech Inc DIY0916U003
Encompass_Preferred
The Rabbit CXCL10 (IP-10) ELISA contains capture antibody standard and detection antibody for development of a Rabbit CXCL10 ELISA. The antibodies have been determined to function in an ELISA with the standard provided. Optimal buffers concentrations incubation times incubation temperatures and methods for the ELISA have not been determined. This allows for complete flexibility to provide optimal dynamic range and sensitivity based on your sample type. Alternate names include CXCL10 C7 IFI10 INP10 IP-10 SCYB10 crg-2 gIP-10 mob-1 C-X-C motif chemokine ligand 10 C-X-C motif chemokine 10. The quantities of components provided are not matched. Components may also be purchased separately. For research use only. Made in the USA
Catalog No. 50-228-5285
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.