Learn More
Rockland ImmunochemicalsSupplier Diversity Partner Antibody for the detection of FLAG™ conjugated proteins (RABBIT) Antibody Biotin Conjugated - 600-406-383 DFS Item
Manufacturer:  Rockland ImmunochemicalsSupplier Diversity Partner 600406383
This antibody is optimally suited for monitoring the expression of FLAG™ tagged fusion proteins.   The antibody recognizes the epitope tag fused to the N or C termini of targeted proteins.  This antibody has been tested by ELISA and WB against both the immunizing peptide and FLAG™ containing recombinant proteins.  This antibody is likely functional for IP and ICC, and other immunodetection techniques.  The epitope tag peptide sequence was first derived from the 11-amino-acid leader peptide of the gene-10 product from bacteriophage T7.  Now the most commonly used hydrophilic octapeptide is DYKDDDDK.  Rockland Immunochemical's antibody to detect FLAG™ conjugated proteins binds FLAG™ containing fusion proteins with greater affinity than the widely used monoclonal M1, M2 and M5 clones, and shows greater sensitivity in most assays. 
Catalog No. NC1479432