Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Rabbit IgG ELISA Kit (Colorimetric)
SDP

Catalog No. NB397811 Shop All R&D Systems Products
Click to view available options
Quantity:
96 wells

Catalog No. NB397811

Supplier: Novus Biologicals™ NBP312526

Only null left
Add to Cart
Add to Cart

Kit (Colorimetric)

Specifications

Assay Range 0.69 to 500 ng/mL
Detection Method ELISA
Immunoassay Kit Format Colorimetric Detection Kit
Product Type ELISA Kit (Colorimetric)
Sample Type Serum, Plasma
Target Species Rabbit
For Use With (Application) ELISA
Gene ID (Entrez) 3500
Gene Symbol IGHG1
Quantity 96 wells
Research Discipline Adaptive Immunity, Epitope Tags, Immunology
Storage Requirements Store at 4°C.
Synonym Immunoglobulin G, ImmunoglobulinG, IP Detection Reagent
Target Rabbit
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.