Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit IgG, F(ab)2, Rhodamine, Isotype control, Novus Biologicals™

Rabbit Isotype Control

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP196892

Catalog No. NBP196892


Add to cart

Description

Description

IgG F(ab)2 Isotype Control antibody specifically detects IgG F(ab)2 in Rabbit samples. It is validated for SDS-Page.
Specifications

Specifications

IgG F(ab)2
LYOPH
SDS-Page 1:100 - 1:2000
Conjugated
IgG
1 mg
Store lyophilized antibody at 4°C. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Rabbit IgG, anti-Rabbit IgG F(ab)2 and anti-Rabbit Serum. No reaction was observed against anti-Rabbit IgG F(c) or anti-Pepsin.
Rabbit
SDS-Page
Rhodamine
0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2. Stabilizer: 10 mg/mL BSA - Immunoglobulin and Protease free. with 0.01% Sodium Azide
Rabbit
Ion exchange chromatography
RUO
Primary
Reconstitute with 1.0 ml deionized water (or equivalent) to desired concentration.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit