Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Rabbit IgG (H+L) Cross-Adsorbed F(ab')2-Goat anti-Rabbit, Alexa Fluor Plus 647, Secondary Antibody, Invitrogen™ Non-distribution product as customer accommodation.

F(ab')2-Goat Polyclonal Antibody

$159.00 - $249.00

Specifications

Antigen Rabbit IgG (H+L) Cross-Adsorbed
Concentration 2 mg/mL
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
Classification Polyclonal
Conjugate Alexa Fluor Plus 647
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
A48285 Non-distribution product as customer accommodation.
View Documents
LIFE TECHNOLOGIES
A48285
500 μg
Each for $249.00
Add to cart
 
A48285TR Non-distribution product as customer accommodation.
View Documents
LIFE TECHNOLOGIES
A48285TR
100 μg
Each for $159.00
Add to cart
 
Description

Description

F(ab')2 antibody fragments offer several benefits over whole IgGs, due to their smaller size (∽110 kD vs. ∽ 150 kD) for better penetration into tissue sections, resulting in improved staining in applications such as IHC. The lack of Fc region results in reduced nonspecific background staining when staining cells with Fc receptors (such as immune cells) and other Fc interaction-related interference. F(ab')2 antibody fragments were obtained by a combination of pepsin digestion and immunoaffinity chromatography to remove residual Fc fragments and undigested whole IgG molecules. To minimize cross-reactivity, the goat anti-rabbit IgG F(ab')2 fragments have been cross-adsorbed against human, mouse and rat serum proteins to increase specificity of the antibody resulting in higher sensitivity and less background staining. This antibody binds to heavy chains on rabbit IgG and light chains on all rabbit immunoglobulins, does not bind to human, mouse or rat serum proteins/IgG or rabbit non-immunoglobulin serum proteins.

Rabbit IgG (H+L) Cross-Adsorbed Polyclonal antibody specifically detects Rabbit IgG (H+L) Cross-Adsorbed in Rabbit samples. It is validated for Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
Specifications

Specifications

Rabbit IgG (H+L) Cross-Adsorbed
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
Alexa Fluor Plus 647
Goat
Rabbit
Gamma Immunoglobins Heavy and Light chains
Polyclonal
Affinity chromatography
Secondary Antibody
2 mg/mL
Polyclonal
Liquid
RUO
Proprietary buffer with 0.016% Bromonitrodioxane, 0.016% Methylisothiazolone; pH 6.5
IgG
Secondary
4° C, store in dark
SDS
Product Certifications
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit