Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Rabbit IgG Recombinant Isotype Control (HyHEL-10)

Catalog No. MA555158
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
Catalog No. Quantity
MA555158 100 μg
1 options

Catalog No. MA555158

Supplier: Invitrogen™ MA555158

Only null left
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Isotype Control

Rabbit IgG Isotype Control for Flow

Rabbit IgG refers to immunoglobulin G antibodies produced by rabbits in response to an antigen. These antibodies are commonly used in laboratory research as a detection tool for specific proteins, molecules, or cells. Rabbit IgG antibodies are highly specific and have a strong affinity for their target, making them ideal for use in various applications such as western blotting, immunohistochemistry, and flow cytometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Rabbit IgG
Applications Flow Cytometry
Classification Recombinant Monoclonal
Clone HyHEL-10
Concentration 2.25 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene Alias Immunoglobulin G; ImmunoglobulinG; Not Applicable
Host Species Rabbit
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Target Species Not Applicable
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Isotype Control
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.