Learn More
Raybiotech IncSupplier Diversity Partner Rabbit IL-2 ELISA Kit DFS Item
Manufacturer:  Raybiotech IncSupplier Diversity Partner ELLIL21
For cell culture supernatants, plasma, and serum samples. Sensitivity: 2.4 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair detects Rabbit IL-2. Other species not determined.
Catalog No. 50-194-9906