Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Novus Biologicals™ Rabbit Insulin ELISA Kit (Colorimetric)

Assay has high sensitivity and specificity for detection of rabbit INS

$702.50

Specifications

Sensitivity 5 uIU/ml
Sample Type Plasma, Serum, Tissue Homogenates
Research Discipline Lipid and Metabolism
Specificity For detection of rabbit INS
Synonym IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB600731KT View Documents Novus Biologicals™
NBP2600731KIT
1 Kit Each for $702.50
Add to cart
 
Description

Description

The Rabbit Insulin ELISA Kit (Colorimetric) measures rabbit Insulin in serum, plasma, tissue homogenates.
Specifications

Specifications

5 uIU/ml
Lipid and Metabolism
IDDM2, ILPR, insulin, IRDN, MODY10, proinsulin
Insulin
3630
ELISA Kit (Colorimetric)
50 to 100 μL
Plasma, Serum, Tissue Homogenates
For detection of rabbit INS
Store the unopened product at 2° to 8°C. Protect from light. Do not use past expiration date.
ELISA
INS
8 to 100uIU/mL
Instruction manual, Standard, Stop Solution, Substrate A, Wash Buffer (20 x concentrate), Substrate B, HRP-conjugate, Adhesive Strip (For 96 wells), Assay plate

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit