Learn More
Novus Biologicals™ Rabbit Lipocalin-2/NGAL ELISA Kit (Colorimetric)
Rabbit Lipocalin-2/NGAL ELISA Kit (Colorimetric) from the most referenced ELISA manufacturer. Highly validated for accurate quantitation and long-term reproducibility.
Manufacturer:  Novus Biologicals™ NBP276788
Catalog No. NBP276788
Add to cart
Description
Description
The Rabbit Lipocalin-2/NGAL ELISA Kit (Colorimetric) measures rabbit Lipocalin-2/NGAL in serum, plasma and other biological fluids.
Specifications
Specifications
78.125 to 5000 pg/mL
ELISA Kit (Colorimetric)
Storage is content dependent.
LCN2
Biological Fluids, Plasma, Serum
3934
46.875 pg/mL
Rabbit
1 Kit