Learn More
Novus Biologicals™ Rabbit Oxidized LDL ELISA Kit (Colorimetric)
This kit recognizes Rabbit OxLDL in samples.
Supplier: Novus Biologicals™ NBP279690
Description
No Significant cross-reactivity or interference between Rabbit OxLDL and analogues was observed.Specifications
0.16 to 10 ng/mL | |
ELISA Kit (Colorimetric) | |
This kit recognizes Rabbit Oxidized LDL in samples. No Significant cross-reactivity or interference between Rabbit Oxidized LDL and analogues was observed. | |
Rabbit | |
Storage of components varies. See protocol for specific instructions. |
0.10 ng/mL | |
Other Biological Fluids, Plasma, Serum | |
ELISA | |
1 Kit | |
Oxidized Low Density Lipoprotein |