Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Our pricing system is unavailable. You are viewing list price.
Please call 1-800-766-7000 to place your order or try our site again later.

Novus Biologicals™ Rabbit Plasminogen ELISA Kit (Colorimetric)

Assay has high sensitivity and specificity for detection of rabbit Plg.

$1004.00

Specifications

Species Rabbit
Sensitivity 15.6ng/mL
Sample Type serum, plasma, tissue homogenates
Research Discipline Cardiovascular Biology
Specificity For detection of rabbit Plg
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB601141KT View Documents Novus Biologicals™
NBP2601141KIT
1 Kit Each for $1,004.00
N/A
Description

Description

The Rabbit Plasminogen ELISA Kit (Colorimetric) measures rabbit Plasminogen in serum, plasma, tissue homogenates.
Specifications

Specifications

Rabbit
serum, plasma, tissue homogenates
For detection of rabbit Plg
Store the unopened product at 2 to 8°C Protect from light. Do not use past expiration date.
ELISA
PLG
62.5ng/mL to 4000ng/mL
Instruction manual, Stop Solution, Sample Diluent, Standard (Freeze dried), TMB Substrate, Wash Buffer (25 x concentrate), HRP-conjugate (100 x concentrate), HRP-conjugate Diluent, Adhesive Strip (For 96 wells), Assay plate (12 x 8 coated Microwells)
15.6ng/mL
Cardiovascular Biology
DKFZp779M0222, EC 3.4.21, EC 3.4.21.7, plasmin, plasminogen
Plasminogen
5340
ELISA Kit (Colorimetric)
50 - 100μL

For Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit