Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ Rabbit Serum

Catalog No. p-2625678
Click to view available options
Quantity:
20 mL
100 mL
Form:
Liquid
Concentration:
4 to 7 g/dL

Sterile, Frozen Brown Solution. Combine with BSA to use as a blocking serum in immunocytochemistry and immunohistochemistry.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance U.S. regulations
Content And Storage -20°C
Species Rabbit
Form Liquid
Product Type Rabbit Serum
Quantity 100 mL
Concentration 4 to 7 g/dL
Sterility Sterile
Sterilization Method Membrane-filtered
For Use With (Application) Blocking agent in immunohistochemistry culture media component
pH 6.5 to 8
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.