Learn More
BioworldSupplier Diversity Partner Rabbit serum, sterile filtered, 0.2 absolute, heat inactivated, 500 mL DFS Item
Manufacturer:  BioworldSupplier Diversity Partner 306111582
Collected from 8-12 week-old New Zealand or Californian rabbits.Aseptically collected and processed to minimize hemolytic activity.Physiochemical constituents are measured to ensure lot-to-lot consistency.Sterility tested.Low Hemoglobin levels.Sterile filtered to 0.2 µm.Storage Conditions: -20°C + 5°C.Usage:bioWORLD's products are supplied for LABORATORY RESEARCH USE ONLY.The product may not be used as a drug,agricultural or pesticidal product,food additive or as a household chemical.
Catalog No. 50-254-237