Learn More
Rabbits, Bio-Fresh™
SCI_ED
Oryctolagus cuniculus.
$57.60
Specifications
Preservative Bio-Fresh™
View More Specs
Products 5
Catalog Number Mfr. No. Injection Type Type Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Injection Type Type Quantity Price Quantity & Availability  
S1851S
SCI_ED
View Documents
Ranaco Corporation
140.1054.1
Double Fur Rabbit 1/Pk.
Pack for $69.60
N/A
Due to product restrictions, please to purchase or view availability for this product.
S1854S
SCI_ED
View Documents
Ranaco Corporation
140.1062.1
Double Fur Rabbit, Pregnant 4/Cs.
Pack for $72.80
N/A
Due to product restrictions, please to purchase or view availability for this product.
S1059S
SCI_ED
View Documents
Ranaco Corporation
140.1059.1
Double Skinned Rabbit Ea.
Pack for $57.60
Add to cart
S1853S
SCI_ED
View Documents
N/A Plain Fur Rabbit 6/Cs. Due to product restrictions, please to purchase or view availability for this product.This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
S1057S
SCI_ED
View Documents
N/A Plain Skinned Rabbit Ea. This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
Description
Description
Excellent for herbivore vs. carnivore comparative studies. Excellent herbivore specimen for study. We recommend dissecting at least one rabbit along with your normal cat study.
Strongest comparisons between the rabbit and cat anatomy exist within:
  • Intestinal systems
  • Eye position and visual acuity
  • Dentition
  • Forearm musculature and functionality
  • Each rabbit comes with a tag and label identifying the sex
Order Info
INJECTION KEY:
Plain: Circulatory system not injected with colors
Single: Arteries injected with red latex
Double: Arteries red, veins blue
Triple: Arteries red, veins blue and hepatic portal veins yellow
Specifications
Specifications
Bio-Fresh™