Description
Excellent for herbivore vs. carnivore comparative studies. Excellent herbivore specimen for study. We recommend dissecting at least one rabbit along with your normal cat study.
Strongest comparisons between the rabbit and cat anatomy exist within:
- Intestinal systems
- Eye position and visual acuity
- Dentition
- Forearm musculature and functionality
- Each rabbit comes with a tag and label identifying the sex
Order Info
INJECTION KEY
Plain: Circulatory system not injected with colors
Single: Arteries injected with red latex
Double: Arteries red, veins blue
Triple: Arteries red, veins blue, and hepatic portal veins yellow
Specifications
Fur Rabbit | |
Fisher-Free™ | |
Plain | |
6/Cs. |