Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RABEP2 Monoclonal Antibody (2B1B6)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA548782
Description
RABEP2 Monoclonal Antibody for Western Blot, IHC (P), Flow, ELISA
RABEP2, also named as RABPT5B, plays a role in membrane trafficking and in homotypic early endosome fusion. It has two isoforms with MW 59-63 kDa.
Specifications
RABEP2 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
Q9H5N1 | |
RABEP2 | |
Purified recombinant fragment of human RABEP2 (AA: 10-200) expressed in HEK293-6e cells supernatant. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
2B1B6 | |
Unconjugated | |
RABEP2 | |
2610011A08Rik; AI462746; fos-related antigen; Fra; MP13; MP13 protein; Rab GTPase-binding effector protein 2; rabaptin, RAB GTPase binding effector protein 2; Rabaptin-5 beta; rabaptin-5beta; RABEP2; RABPT5B | |
Mouse | |
Protein G | |
RUO | |
79874 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction