Learn More
RABEPK/p40 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 151051AP
Catalog No. 50-172-9982
Add to cart
Description
Description
The protein encoded by this gene is a cytokine that acts as a regulator of a variety of hematopoietic cells. This cytokine stimulates cell proliferation and prevents apoptosis. It functions through the interleukin 9 receptor (IL9R), which activates different signal transducer and activator (STAT) proteins and thus connects this cytokine to various biological processes. The gene encoding this cytokine has been identified as a candidate gene for asthma. Genetic studies on a mouse model of asthma demonstrated that this cytokine is a determining factor in the pathogenesis of bronchial hyperresponsiveness.
Specifications
Specifications
RABEPK/p40
0.18 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q7Z6M1
40 kDa Rab9 effector protein, p40, RAB9P40, RABEPK
Rabbit
IgG
150 μL
-20°C
Primary
10244
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
RABEPK
Liquid
RABEPK
RABEPK/p40 Fusion Protein Ag2593
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human