Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ RABGGTB Recombinant Protein
$503.00 - $775.00
Specifications
Accession Number | AAH20790 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer |
Gene ID (Entrez) | 5876 |
Molecular Weight (g/mol) | 61.93 |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
89-010-450
|
Abnova™
H00005876P0110 |
10 ug |
Each for $503.00
|
|
|||||
89-010-451
|
Abnova™
H00005876P0125 |
25 ug |
Each for $775.00
|
|
|||||
Description
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
AAH20790 | |
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer | |
61.93 | |
8 | |
Glutathione Sepharose 4 Fast Flow | |
Wheat Germ (in vitro) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
RABGGTB | |
Human | |
Recombinant | |
Solution |
Antibody Production, Protein Array, ELISA, Western Blot | |
5876 | |
RABGGTB (Human) Recombinant Protein (P01) | |
In vitro wheat germ expression system | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MGTPQKDVIIKSDAPDTLLLEKHADYIASYGSKKDDYEYCMSEYLRMSGIYWGLTVMDLMGQLHRMNREEILAFIKSCQHECGGISASIGHDPHLLYTLSAVQILTLYDSINVIDVNKVVEYVKGLQKEDGSFAGDIWGEIDTRFSFCAVATLALLGKLDAINVEKAIEFVLSCMNFDGGFGCRPGSESHAGQIYCCTGFLAITSQLHQVNSDLLGWWLCERQLPSGGLNGRPEKLPDVCYSWWVLASLKIIGRLHWIDREKLRNFILACQDEETGGFADRPGDMVDPFHTLFGIAGLSLLGEEQIKPVNPVFCMPEEVLQRVNVQPELVS | |
GGTB | |
RABGGTB | |
Wheat Germ (in vitro) | |
GST |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title