Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RABV Glycoprotein Mouse anti-RabiesVirus, Unconjugated, Clone: MAb-D1, MilliporeSigma™

Mouse Monoclonal Antibody

$147.00 - $372.00

Specifications

Antigen RABV Glycoprotein
Applications ELISA, Immunoprecipitation, Neutralization
Conjugate Unconjugated
Format Biotin
Host Species Mouse
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
MABF19625MI
View Documents
MilliporeSigma
MABF1967B25UG
25 μg
Each for $147.00
Add to cart
 
MABF1967BMI
View Documents
MilliporeSigma
MABF1967B100UG
100 μg
Each for $372.00
Add to cart
 
Description

Description

RABV Glycoprotein Monoclonal antibody specifically detects RABV Glycoprotein in RabiesVirus samples. It is validated for ELISA, Immunoprecipitation, Neutralization Assay.
Specifications

Specifications

RABV Glycoprotein
Unconjugated
Mouse
Glycoprotein
Inactivated and purified Rabies Virus (Pasteur virus strain).
Primary
Stable for 1 year at 2 to 8°C from date of receipt.
ELISA, Immunoprecipitation, Neutralization
Biotin
Rabies Virus
G
Monoclonal
Protein G Purified
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit