Learn More
Medchemexpress LLC (Rac)-EBET-1055 | 3031540-65-2 | 97.2% | 923.02 | 25 MG
SDP
Supplier:  Medchemexpress LLC 25MGHY161346A
Encompass_Preferred
(Rac)-EBET-1055 is the racemate of EBET-1055. It is a bromodomain and extra-terminal (BET) PROTAC degrader that effectively inhibits the growth of pancreatic ductal adenocarcinoma (PDAC) and simultaneously modulates cancer-associated fibroblast (CAF) activity, upregulating all reporter gene activities in organoid co-cultures.
  • Racemate of EBET-1055
  • Bromodomain and extra-terminal (BET) PROTAC degrader
  • Inhibits the growth of pancreatic ductal adenocarcinoma (PDAC)
  • Modulates cancer-associated fibroblast (CAF) activity
  • Upregulates reporter gene activities in organoid co-cultures
Catalog No. 50-003-91630