Learn More
Cell Biolabs IncSupplier Diversity Partner RAC1 ACTIVATION ASSAY KIT DFS Item
Manufacturer:  Cell Biolabs IncSupplier Diversity Partner STA4011
The Rac1 Activation Assay Kit provides a non-radioactive assay format to pull down the active form of Rac with colored agarose beads, which allow for a visual check, followed by Western blot with an Anti-Rac1 antibody. 20 assays/kit.
Catalog No. NC0045609