Learn More
Cell Biolabs Inc
RAC1 ELISA 96W COLORIMETRIC
Manufacturer: Cell Biolabs Inc STA440
The 96-Well Ras Activation ELISA Kit uses the Raf1 RBD bound to a 96-well plate to pull down only the active form of Ras. The captured GTP-Ras is then detected by colorimetric plate reader. 96 wells/kit.