Sino Biological™ RAC1 Recombinant Human Protein, N-GST Tag Non-distribution product as customer accommodation.

Expressed from DNA sequence encoding mature form of human Rac1 isoform A (Met 1-Cys 189) fused to GST tag at N-terminus

Manufacturer: Sino Biological™ 10535H09B5

 View more versions of this product

Catalog No. 10-535-H09B5

  Check Availability
Add to Cart

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


    Specifications

    Structural Form Full Length, Recombinant
    Molecular Weight 47kd
    Quantity 5μg
    Common Name Ras-Related C3 Botulinum Toxin Substrate 1 Protein
    Gene Symbol RAC 1
    Expression System Insect
    Purity ≥90% by SDS-PAGE
    Source Human
    Endotoxin Concentration <1 EU/μg
    Protein Tag N-GST Tag
    Validated Application Assay Standard
    Protein Family Other proteins
    Product Type RAC1 Recombinant Human Protein
    For Use With (Application) Analysis of protein structure, used as immunogen, protein standard, or in cell biology research applications
    Format Lyophilized
    Gene ID (Entrez) 5879

    Expressed from DNA sequence encoding mature form of human Rac1 isoform A (Met 1-Cys 189) fused to GST tag at N-terminus

    • Formulation: 50mM Tris, 100mM NaCl, 1mM GSH, 10% Glycerol, pH 8.0.

  • Description & Specifications

Related Products


Product Content Feedback

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit